Gene Transcripts
- Gene Symbol
- Eya2
- Entrez Gene ID
- 14049
- Gene Name
- eyes absent 2 homolog (Drosophila)
- Gene Aliases
- Eab1
- Chromosome Location
- Chr. 2: 165595028 - 165771727 on Build GRCm38
- UniGene ID
- Mm.282719
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001271962.1 |
NP_001258891.1
|
11, 12 |
1101 |
|
NM_001271963.1 |
NP_001258892.1
|
11, 12 |
1277 |
|
NM_010165.3 |
NP_034295.1
|
11, 12 |
1278 |
|
GenBank mRNA |
AK137718.1 |
- |
11, 12 |
1273 |
|
AK141765.1 |
- |
11, 12 |
1277 |
|
AY212511.1 |
- |
8, 9 |
792 |
|
AY212512.1 |
- |
9, 10 |
948 |
|
BC003755.1 |
- |
11, 12 |
1278 |
3497245 |
U61111.1 |
- |
10, 11 |
1019 |
|
U71208.1 |
- |
11, 12 |
1103 |
|
U81603.1 |
- |
10, 11 |
1018 |
|
View Full Product Details