Gene Transcripts
- Gene Symbol
- Fxn
- Entrez Gene ID
- 14297
- Gene Name
- frataxin
- Gene Aliases
- FA, FARR, Frda, X25
- Chromosome Location
- Chr. 19: 24261453 - 24280586 on Build GRCm38
- UniGene ID
- Mm.7319
- Species
- Mus musculus
- Gene Symbol
- Gm5037
- Entrez Gene ID
- 260347
- Gene Name
- frataxin pseudogene
- Gene Aliases
- EG260347, Frda-ps
- Chromosome Location
- Chr. 7: 95912745 - 95913390 on Build GRCm38
- UniGene ID
- Mm.439173
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AF223569.1 |
- |
1 |
256 |
|
View Full Product Details