Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001271981.1 | NP_001258910.1 | 2 | 554 | |
NM_008051.6 | NP_032077.2 | 2 | 692 | ||
GenBank mRNA | AF113533.1 | - | 1 | 548 | |
AK137638.1 | - | 2 | 694 | ||
BC109145.1 | - | 2 | 530 | 40045410 | |
BC109146.1 | - | 2 | 530 | 40045419 |