Gene Transcripts

Gene Symbol
PSMB7
Entrez Gene ID
5695
Gene Name
proteasome subunit beta 7
Gene Aliases
Z
Chromosome Location
Chr. 9: 124353465 - 124415473 on Build GRCh38
UniGene ID
Hs.213470
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002799.3 NP_002790.1 3 290
GenBank mRNA AK098757.1 - 3 259
AK223007.1 - 3 243
AK223356.1 - 3 255
AK303460.1 - 3 259
AK308756.1 - 3 290
AK314302.1 - 3 259
BC000509.2 - 3 246 2822305
BC008414.1 - 3 256
BC017116.2 - 2 181
BT007218.1 - 3 242
CR456721.1 - 3 242
D38048.1 - 3 256
GU727627.1 - 3 259
U44425.1 - 3 230

View Full Product Details