Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001010941.2 | NP_001010941.1 | 2 | 971 | |
NM_008151.3 | NP_032177.1 | 1 | 757 | ||
GenBank mRNA | AK140298.1 | - | 2 | 972 | |
BC055746.1 | - | 1 | 757 | 6415225 | |
D21061.1 | - | 2 | 559 |