Gene Transcripts

Gene Symbol
HNRNPH3
Entrez Gene ID
3189
Gene Name
heterogeneous nuclear ribonucleoprotein H3
Gene Aliases
2H9, HNRPH3
Chromosome Location
Chr. 10: 68331174 - 68343196 on Build GRCh38
UniGene ID
Hs.643472
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001322434.1 NP_001309363.1 3 390
NM_001322436.1 NP_001309365.1 3 352
NM_001322437.1 NP_001309366.1 3 455
NM_001322438.1 NP_001309367.1 3 327
NM_001322439.1 NP_001309368.1 3 390
NM_001322440.1 NP_001309369.1 3 352
NM_001322441.1 NP_001309370.1 3 455
NM_001322442.1 NP_001309371.1 3 390
NM_001322443.1 NP_001309372.1 3 371
NM_012207.2 NP_036339.1 3 371
NM_021644.3 NP_067676.2 3 371
XM_017016168.1 XP_016871657.1 3 801
GenBank mRNA AF132361.1 - 3 260
AF132362.1 - 1 272
AK091411.1 - 2 1737
AK223441.1 - 3 323
AK291547.1 - 3 323
AL117395.1 - 1 1087
BC039824.1 - 3 307 6061550

View Full Product Details