Gene Transcripts

Gene Symbol
TLX1
Entrez Gene ID
3195
Gene Name
T-cell leukemia homeobox 1
Gene Aliases
HOX11, TCL3
Chromosome Location
Chr. 10: 101130773 - 101137789 on Build GRCh38
UniGene ID
Hs.89583
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001195517.1 NP_001182446.1 3 1212
NM_005521.3 NP_005512.1 3 1189
XM_011539744.2 XP_011538046.1 3 1741
GenBank mRNA BC130530.1 - 3 1212
M62626.1 - 3 1969
M75952.1 - 3 963
S38742.1 - 3 1049

View Full Product Details