Gene Transcripts

Gene Symbol
Hic1
Entrez Gene ID
15248
Gene Name
hypermethylated in cancer 1
Gene Aliases
AA408311, HIC-1
Chromosome Location
Chr. 11: 75164565 - 75170255 on Build GRCm38
UniGene ID
Mm.57250
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001098203.1 NP_001091673.1 2 472
NM_010430.2 NP_034560.2 2 1466
GenBank mRNA CJ067923.1 - 2 356

View Full Product Details