Gene Transcripts

Gene Symbol
Hk2
Entrez Gene ID
15277
Gene Name
hexokinase 2
Gene Aliases
AI642394, HKII
Chromosome Location
Chr. 6: 82725025 - 82774454 on Build GRCm38
UniGene ID
Mm.255848
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013820.3 NP_038848.1 16 2746
GenBank mRNA AJ238540.1 - 16 2689
AK145936.1 - 16 2727
BC054472.1 - 16 2665 6465367

View Full Product Details