Gene Transcripts

Gene Symbol
Il1a
Entrez Gene ID
16175
Gene Name
interleukin 1 alpha
Gene Aliases
Il-1a
Chromosome Location
Chr. 2: 129299609 - 129310901 on Build GRCm38
UniGene ID
Mm.15534
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010554.4 NP_034684.2 4 203
GenBank mRNA AK156449.1 - 4 204
X01450.1 - 4 203

View Full Product Details