Gene Transcripts

Gene Symbol
Mapkapk5
Entrez Gene ID
17165
Gene Name
MAP kinase-activated protein kinase 5
Gene Aliases
MK5, PRAK
Chromosome Location
Chr. 5: 121525051 - 121545892 on Build GRCm38
UniGene ID
Mm.272206
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010765.2 NP_034895.1 4 922
GenBank mRNA AF039840.1 - 4 936
AY533679.1 - 4 230
AY533680.2 - 4 230
BC019184.1 - 4 344 5010305

View Full Product Details