Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_013594.2 | NP_038622.2 | 9, 10 | 1161 | |
GenBank mRNA | AF072240.1 | - | 10, 11 | 1140 | |
AK004624.2 | - | 9, 10 | 1162 | ||
AK032392.1 | - | 1, 2 | 105 | ||
AK032535.1 | - | 6, 7 | 659 | ||
AK156401.1 | - | 10, 11 | 1152 | ||
AK166042.1 | - | 5, 6 | 481 | ||
BC069837.1 | - | 9, 10 | 1177 | 30435760 |