Gene Transcripts

Gene Symbol
Myo15
Entrez Gene ID
17910
Gene Name
myosin XV
Gene Aliases
Myo15a, sh-2, sh2
Chromosome Location
Chr. 11: 60469339 - 60528369 on Build GRCm38
UniGene ID
Mm.89960
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001103171.1 NP_001096641.1 18 5309
NM_010862.2 NP_034992.2 18 5309
NM_182698.2 NP_874357.2 17 1547
GenBank mRNA AB014510.1 - 14 1312
AF053130.1 - 16 1515
AF144095.1 - 18 5309
AY331132.1 - 18 5309
AY331133.1 - 17 1547

View Full Product Details