Gene Transcripts

Gene Symbol
Nek1
Entrez Gene ID
18004
Gene Name
NIMA (never in mitosis gene a)-related expressed kinase 1
Gene Aliases
D8Ertd790e, kat
Chromosome Location
Chr. 8: 60993193 - 61131349 on Build GRCm38
UniGene ID
Mm.486881
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001293637.1 NP_001280566.1 6 1053
NM_001293638.1 NP_001280567.1 6 1053
NM_001293639.1 NP_001280568.1 6 1053
NM_175089.4 NP_780298.2 6 1053
GenBank mRNA AK045202.1 - 5 909
AK077047.1 - 7 724
AK173292.1 - 6 1051
AY850065.1 - 6 1026
BC070443.1 - 6 996
BC157939.1 - 6 981 9088119
BC158009.1 - 6 981 9007317
BC172091.1 - 6 981 9054377

View Full Product Details