Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_010946.3 | NP_035076.1 | 7, 8 | 623 | |
GenBank mRNA | AK041112.1 | - | 7, 8 | 580 | |
AK214245.1 | - | 5, 6 | 371 | ||
BC030172.1 | - | 7, 8 | 621 | 5037501 | |
U57692.1 | - | 7, 8 | 577 |