Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_010950.2 | NP_035080.2 | 10 | 2636 | |
GenBank mRNA | AK147523.1 | - | 8 | 8423 | |
BC059915.1 | - | 9, 10 | 3176 | ||
BC068116.1 | - | 10 | 2463 | 6809443 | |
BC098097.1 | - | 10 | 2477 | 6403012 | |
U96441.1 | - | 11 | 2512 |