Gene Transcripts

Gene Symbol
Orm1
Entrez Gene ID
18405
Gene Name
orosomucoid 1
Gene Aliases
Agp-1, Agp-2, Orm-1
Chromosome Location
Chr. 4: 63344566 - 63348162 on Build GRCm38
UniGene ID
Mm.4777
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008768.1 NP_032794.1 2 225
GenBank mRNA BC012725.1 - 2 232
CT010399.1 - 2 193
M27008.1 - 2 225

View Full Product Details