Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_008774.3 | NP_032800.2 | 2 | 815 | |
GenBank mRNA | AK005009.1 | - | 2 | 816 | |
AK044829.1 | - | 2 | 812 | ||
AK089747.1 | - | 2 | 815 | ||
AK133196.1 | - | 2 | 816 | ||
AK150839.1 | - | 2 | 815 | ||
AK152064.1 | - | 2 | 783 | ||
AK159703.1 | - | 2 | 815 | ||
AK159733.1 | - | 2 | 815 | ||
AK160883.1 | - | 2 | 815 | ||
AK160968.1 | - | 2 | 814 | ||
AK161123.1 | - | 2 | 818 | ||
AK161671.1 | - | 2 | 814 | ||
AK168466.1 | - | 2 | 814 | ||
BC003870.1 | - | 2 | 434 | 3491915 | |
BC004587.1 | - | 1 | 21 | ||
BC011207.1 | - | 2 | 572 | 4022169 | |
BC023145.1 | - | 2 | 410 | 3988101 | |
BC046233.1 | - | 2 | 805 | 6816124 | |
X65553.1 | - | 2 | 601 |