Gene Transcripts
- Gene Symbol
- IFI16
- Entrez Gene ID
- 3428
- Gene Name
- interferon gamma inducible protein 16
- Gene Aliases
- IFNGIP1, PYHIN2
- Chromosome Location
- Chr. 1: 158999968 - 159055155 on Build GRCh38
- UniGene ID
- Hs.380250
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001206567.1 |
NP_001193496.1
|
8, 9 |
1779 |
|
NM_005531.2 |
NP_005522.2
|
8, 9 |
1779 |
|
XM_005245127.3 |
XP_005245184.1
|
10, 11 |
2264 |
|
GenBank mRNA |
AB208989.1 |
- |
8, 9 |
1749 |
|
AF208043.1 |
- |
8, 9 |
1753 |
|
AK296228.1 |
- |
8, 9 |
1662 |
|
AK307957.1 |
- |
8, 9 |
1560 |
|
AY138863.1 |
- |
7, 8 |
1489 |
|
BC017059.1 |
- |
8, 9 |
1746 |
3914632 |
DA945454.1 |
- |
3, 4 |
351 |
|
M63838.1 |
- |
8, 9 |
1753 |
|
View Full Product Details