Gene Transcripts

Gene Symbol
Psme3
Entrez Gene ID
19192
Gene Name
proteaseome (prosome, macropain) activator subunit 3 (PA28 gamma, Ki)
Gene Aliases
AA410043, AU020960, Ki, PA28gamma, REGgamma, pa28g
Chromosome Location
Chr. 11: 101316251 - 101323531 on Build GRCm38
UniGene ID
Mm.288477
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011192.3 NP_035322.1 6 553
GenBank mRNA AK140597.1 - 6 574
AK150939.1 - 6 573
AK156079.1 - 6 523
AK171348.1 - 6 569
AK178475.1 - 5 274
AK182764.1 - 4 297
AK183517.1 - 4 297
AK183726.1 - 4 261
AK184446.1 - 4 297
AK196904.1 - 4 296
AK198462.1 - 4 297
AK210357.1 - 4 293
BC015255.1 - 6 553 4221225
BC139818.1 - 6 501 40141268
CT010198.1 - 6 357
D87911.1 - 6 476

View Full Product Details