Gene Transcripts

Gene Symbol
Ptprcap
Entrez Gene ID
19265
Gene Name
protein tyrosine phosphatase, receptor type, C polypeptide-associated protein
Gene Aliases
AI605264, CD-45-AP, LPAP, LSM-1
Chromosome Location
Chr. 19: 4154646 - 4156710 on Build GRCm38
UniGene ID
Mm.271937
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016933.3 NP_058629.2 2 699
GenBank mRNA AI627045.1 - 1 334
AK088334.1 - 2 697
AK165203.1 - 2 697
BC013273.1 - 2 698 4208992
BC100560.1 - 2 680 832557
D10105.1 - 2 675
U03856.1 - 1 692

View Full Product Details