Gene Transcripts

Gene Symbol
Rab4b
Entrez Gene ID
19342
Gene Name
RAB4B, member RAS oncogene family
Gene Aliases
1500031G17Rik
Chromosome Location
Chr. 7: 27168424 - 27178901 on Build GRCm38
UniGene ID
Mm.482114
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029391.2 NP_083667.1 4, 5 425
GenBank mRNA AB232592.1 - 4, 5 284
AF408432.1 - 4, 5 397
AK005314.1 - 4, 5 425
AK140513.1 - 3, 4 622
AK170552.1 - 4, 5 439
BC007147.1 - 4, 5 369 3600193
BC096654.1 - 4, 5 408 5056766

View Full Product Details