Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000575.4 | NP_000566.3 | 6 | 1473 | |
GenBank mRNA | AK314850.1 | - | 6 | 568 | |
BC013142.2 | - | 5 | 518 | 3875617 | |
BT007014.1 | - | 5 | 509 | ||
CR457414.1 | - | 5 | 509 | ||
M15329.1 | - | 5 | 514 | ||
X02531.1 | - | 5 | 509 | ||
X02851.1 | - | 6 | 567 | ||
X56086.1 | - | 6 | 535 |