Gene Transcripts

Gene Symbol
IL1A
Entrez Gene ID
3552
Gene Name
interleukin 1 alpha
Gene Aliases
IL-1A, IL1, IL1-ALPHA, IL1F1
Chromosome Location
Chr. 2: 112773915 - 112785398 on Build GRCh38
UniGene ID
Hs.1722
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000575.4 NP_000566.3 4 1134
GenBank mRNA AK314850.1 - 4 229
BC013142.2 - 3 179 3875617
BT007014.1 - 3 170
CR457414.1 - 3 170
M15329.1 - 3 175
X02531.1 - 3 170
X02851.1 - 4 228
X56086.1 - 4 196

View Full Product Details