Gene Transcripts

Gene Symbol
Rbbp4
Entrez Gene ID
19646
Gene Name
retinoblastoma binding protein 4
Gene Aliases
RBAP48, mRbAp48
Chromosome Location
Chr. 4: 129307100 - 129335370 on Build GRCm38
UniGene ID
Mm.12145
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009030.3 NP_033056.2 7 911
GenBank mRNA AK082419.1 - 7 883
AK166437.1 - 7 882
BC138568.1 - 7 889 8861588
BC138570.1 - 7 890 8861590
U35141.1 - 7 881

View Full Product Details