Gene Transcripts

Gene Symbol
Ccl12
Entrez Gene ID
20293
Gene Name
chemokine (C-C motif) ligand 12
Gene Aliases
MCP-5, Scya12
Chromosome Location
Chr. 11: 82101845 - 82103399 on Build GRCm38
UniGene ID
Mm.867
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011331.2 NP_035461.2 3 261
GenBank mRNA AF065934.1 - 3 227
AF065935.1 - 3 227
AF065936.1 - 3 227
AF065937.1 - 3 227
AF065938.1 - 3 227
AK012356.1 - 3 261
BC027520.1 - 3 216 1548072
U50712.1 - 3 260
U66670.1 - 3 237

View Full Product Details