Gene Transcripts

Gene Symbol
Sh3bp1
Entrez Gene ID
20401
Gene Name
SH3-domain binding protein 1
Gene Aliases
3BP-1
Chromosome Location
Chr. 15: 78899780 - 78912052 on Build GRCm38
UniGene ID
Mm.4462
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009164.2 NP_033190.2 10 974
GenBank mRNA AK145156.1 - 10 914
AK161215.1 - 9 898
AK166337.1 - 10 967
AK172060.1 - 10 972
BC004598.1 - 8 695 3584958
X87671.1 - 7 830

View Full Product Details