Gene Transcripts

Gene Symbol
Slfn2
Entrez Gene ID
20556
Gene Name
schlafen 2
Gene Aliases
Shlf2
Chromosome Location
Chr. 11: 83065112 - 83070678 on Build GRCm38
UniGene ID
Mm.278689
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011408.1 NP_035538.1 2 845
GenBank mRNA AF099973.1 - 2 845
BC138749.1 - 2 800 8861771
BC138750.1 - 2 800 8861772

View Full Product Details