Gene Transcripts
- Gene Symbol
- Sod2
- Entrez Gene ID
- 20656
- Gene Name
- superoxide dismutase 2, mitochondrial
- Gene Aliases
- MnSOD, Sod-2
- Chromosome Location
- Chr. 17: 13007839 - 13018119 on Build GRCm38
- UniGene ID
- Mm.290876
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_013671.3 |
NP_038699.2
|
2, 3 |
439 |
|
GenBank mRNA |
AK002428.1 |
- |
2, 3 |
299 |
|
AK002534.1 |
- |
2, 3 |
300 |
|
AK012354.1 |
- |
2, 3 |
297 |
|
AK135435.1 |
- |
2, 3 |
428 |
|
AK152047.1 |
- |
2, 3 |
297 |
|
AK168082.1 |
- |
2, 3 |
300 |
|
AK168591.1 |
- |
2, 3 |
431 |
|
AK179113.1 |
- |
1, 2 |
59 |
|
AK180974.1 |
- |
1, 2 |
59 |
|
BC010548.1 |
- |
2, 3 |
275 |
3581909 |
BC018173.1 |
- |
1, 2 |
220 |
|
CT010273.1 |
- |
2, 3 |
232 |
|
X04972.1 |
- |
2, 3 |
284 |
|
Z18857.1 |
- |
2, 3 |
267 |
|
View Full Product Details