Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_009373.3 | NP_033399.1 | 4 | 570 | |
GenBank mRNA | AF076928.1 | - | 3 | 485 | |
AF114266.1 | - | 4 | 535 | ||
AK052912.1 | - | 4 | 545 | ||
AK054108.1 | - | 4 | 545 | ||
AK080224.1 | - | 4 | 570 | ||
AK080593.1 | - | 4 | 545 | ||
AK086001.1 | - | 4 | 545 | ||
AK089481.1 | - | 4 | 573 | ||
AK143712.1 | - | 4 | 606 | ||
AK151776.1 | - | 4 | 571 | ||
AK152152.1 | - | 4 | 545 | ||
AK152627.1 | - | 4 | 571 | ||
AK159255.1 | - | 4 | 545 | ||
AK166302.1 | - | 4 | 545 | ||
AK168990.1 | - | 4 | 546 | ||
AK169356.1 | - | 4 | 573 | ||
BC016492.1 | - | 4 | 511 | 3256943 | |
M55154.1 | - | 4 | 507 |