Gene Transcripts

Gene Symbol
Tgm2
Entrez Gene ID
21817
Gene Name
transglutaminase 2, C polypeptide
Gene Aliases
G[a]h, TG2, TGase2, tTG, tTGas
Chromosome Location
Chr. 2: 158116405 - 158146392 on Build GRCm38
UniGene ID
Mm.330731
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009373.3 NP_033399.1 4 608
GenBank mRNA AF076928.1 - 3 523
AF114266.1 - 4 573
AK052912.1 - 4 583
AK054108.1 - 4 583
AK080224.1 - 4 608
AK080593.1 - 4 583
AK086001.1 - 4 583
AK089481.1 - 4 611
AK143712.1 - 4 644
AK151776.1 - 4 609
AK152152.1 - 4 583
AK152627.1 - 4 609
AK159255.1 - 4 583
AK168990.1 - 4 584
AK169356.1 - 4 611
BC016492.1 - 4 549 3256943
M55154.1 - 4 545

View Full Product Details