Gene Transcripts

Gene Symbol
Neu2
Entrez Gene ID
23956
Gene Name
neuraminidase 2
Gene Aliases
MBS, MSS, MTS
Chromosome Location
Chr. 1: 87574027 - 87597845 on Build GRCm38
UniGene ID
Mm.143717
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001160163.1 NP_001153635.1 3 366
NM_001160164.1 NP_001153636.1 2 264
NM_001160165.1 NP_001153637.1 2 392
NM_015750.3 NP_056565.1 2 232
GenBank mRNA AB028023.1 - 2 234
AB048604.1 - 2 489
AF139059.1 - 2 346
AK009828.1 - 1 446
AK145284.1 - 2 265
BC026408.1 - 1 20
BC105657.1 - 2 234 40046586
BC105658.1 - 2 234 40046587
BC105659.1 - 2 234 40046588
BC105661.1 - 1 494 40046584
FJ613504.1 - 3 349

View Full Product Details