Gene Transcripts

Gene Symbol
Srp54a
Entrez Gene ID
24067
Gene Name
signal recognition particle 54A
Gene Aliases
54kDa, Srp54
Chromosome Location
Chr. 12: 55080496 - 55115367 on Build GRCm38
UniGene ID
Mm.471968
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011899.4 NP_036029.2 2 318
GenBank mRNA AK011928.1 - 2 282
BC019683.1 - 2 247 3708250

Gene Symbol
Srp54b
Entrez Gene ID
665155
Gene Name
signal recognition particle 54B
Gene Aliases
Chromosome Location
Chr. 12: 55155104 - 55189213 on Build GRCm38
UniGene ID
Mm.12848
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001100109.1 NP_001093579.1 2 554
GenBank mRNA AK010716.1 - 2 254
AK136033.1 - 2 555
X16319.1 - 283

Gene Symbol
Srp54c
Entrez Gene ID
100101806
Gene Name
signal recognition particle 54C
Gene Aliases
E330029E12Rik
Chromosome Location
Chr. 12: 55230105 - 55279334 on Build GRCm38
UniGene ID
Mm.471968
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001100110.1 NP_001093580.1 244
GenBank mRNA AK090018.1 - 247
AK146826.1 - 282
BC005543.1 - 261 3585216

View Full Product Details