Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_011936.2 | NP_036066.2 | 3 | 191 | |
GenBank mRNA | AJ237917.1 | - | 3 | 177 | |
AK036677.1 | - | 3 | 181 | ||
AK040866.1 | - | 3 | 169 | ||
AK045465.1 | - | 3 | 168 | ||
AK046935.1 | - | 3 | 150 | ||
AK049502.1 | - | 3 | 184 | ||
AK088881.1 | - | 3 | 177 | ||
AK129437.1 | - | 3 | 158 | ||
AK161060.1 | - | 3 | 150 | ||
BC022222.1 | - | 3 | 142 | 4237261 | |
BC057008.1 | - | 3 | 157 | 5708558 | |
BY290198.1 | - | 3 | 192 |