Gene Transcripts

Gene Symbol
KCNC3
Entrez Gene ID
3748
Gene Name
potassium voltage-gated channel subfamily C member 3
Gene Aliases
KSHIIID, KV3.3, SCA13
Chromosome Location
Chr. 19: 50311942 - 50333515 on Build GRCh38
UniGene ID
Hs.467146
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_004977.2 NP_004968.2 2 1418
XM_006723203.2 XP_006723266.1 2 1123
XM_011526925.2 XP_011525227.1 2 1123
XM_011526926.1 XP_011525228.1 2 1123
XM_011526928.2 XP_011525230.1 2 1123
GenBank mRNA AB208930.1 - 1 224
AF055989.1 - 2 1418

View Full Product Details