Gene Transcripts

Gene Symbol
Msrb1
Entrez Gene ID
27361
Gene Name
methionine sulfoxide reductase B1
Gene Aliases
D17Wsu82e, SELX, SelR, Sepr, Sepx1
Chromosome Location
Chr. 17: 24736642 - 24742778 on Build GRCm38
UniGene ID
Mm.28212
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013759.2 NP_038787.1 2 235
GenBank mRNA AF195142.1 - 2 210
AK002339.1 - 2 195
AK002652.1 - 2 194
AK003112.1 - 2 195
AK003624.1 - 2 195
BC090646.1 - 2 235 30946285
BC141146.1 - 2 199 9055758
CX566836.1 - 2 235

View Full Product Details