Gene Transcripts

Gene Symbol
Pdhx
Entrez Gene ID
27402
Gene Name
pyruvate dehydrogenase complex, component X
Gene Aliases
AI481367, E3bp, Pdx1
Chromosome Location
Chr. 2: 103021055 - 103073513 on Build GRCm38
UniGene ID
Mm.315011
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175094.5 NP_780303.1 3 374
GenBank mRNA AK046020.1 - 3 323
AK047670.1 - 3 372
AK050433.1 - 3 375
AK083170.1 - 3 338
AK166257.1 - 3 337
BC061231.1 - 3 335 6532760

View Full Product Details