Gene Transcripts

Gene Symbol
Fbxl12
Entrez Gene ID
30843
Gene Name
F-box and leucine-rich repeat protein 12
Gene Aliases
3110048D16Rik, Fbl12
Chromosome Location
Chr. 9: 20637749 - 20644767 on Build GRCm38
UniGene ID
Mm.24608
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001002846.2 NP_001002846.1 3 981
NM_001286529.1 NP_001273458.1 3 603
NM_001286530.1 NP_001273459.1 4 707
NM_013911.3 NP_038939.2 3 1011
GenBank mRNA AF176525.1 - 3 893
AF313405.1 - 3 499
AK011081.1 - 3 966
AK012811.1 - 3 505
AK030910.1 - 3 981
AK031262.1 - 2 3067
AK137282.1 - 4 2678
DN176232.1 - 4 698

View Full Product Details