Gene Transcripts

Gene Symbol
Spag6
Entrez Gene ID
50525
Gene Name
sperm associated antigen 6
Gene Aliases
PF16
Chromosome Location
Chr. 16: 16753016 - 16829363 on Build GRCm38
UniGene ID
Mm.31701
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015773.2 NP_056588.1 2, 3 273
GenBank mRNA AF173866.1 - 2, 3 210
AK005732.1 - 2, 3 221
AK054199.1 - 1, 2 309
BC090660.1 - 2, 3 216 30673350
BC100455.1 - 2, 3 223 30919921
CJ060006.1 - 2, 3 273

View Full Product Details