Gene Transcripts

Gene Symbol
Hist1h1c
Entrez Gene ID
50708
Gene Name
histone cluster 1, H1c
Gene Aliases
0610008C09Rik, H1.2, H1c, H1f2, H1var1, His1a
Chromosome Location
Chr. 13: 23738807 - 23739531 on Build GRCm38
UniGene ID
Mm.193539
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK002321.1 - 1451
AK135614.1 - 1185
AK155502.1 - 1455
BC089604.1 - 1453 6763721

View Full Product Details