Gene Transcripts

Gene Symbol
Mtag2
Entrez Gene ID
50994
Gene Name
metastasis associated gene 2
Gene Aliases
MAG
Chromosome Location
Chr. 7: 45366163 - 45367948 on Build GRCm38
UniGene ID
Mm.389897
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA BC107317.1 - 1 114
BC107318.1 - 1 114
BC119507.1 - 1 114
BC119508.1 - 1 114
BC127168.1 - 1 114
BC127169.1 - 1 114
U88401.1 - 1 114

View Full Product Details