Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_144523.2 | NP_653106.1 | 6 | 1563 | |
GenBank mRNA | AK004048.1 | - | 2 | 1291 | |
AK031232.1 | - | 6 | 1563 | ||
AK149106.1 | - | 6 | 1495 | ||
AK154951.1 | - | 6 | 1489 | ||
BC006964.1 | - | 6 | 1468 | 3153893 |