Gene Transcripts

Gene Symbol
KPNA1
Entrez Gene ID
3836
Gene Name
karyopherin subunit alpha 1
Gene Aliases
IPOA5, NPI-1, RCH2, SRP1
Chromosome Location
Chr. 3: 122421901 - 122514939 on Build GRCh38
UniGene ID
Hs.161008
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002264.3 NP_002255.3 11 1268
NR_026698.1 12 1391
XM_005247437.3 XP_005247494.1 11 1150
XM_005247439.3 XP_005247496.1 9 1898
XM_011512795.2 XP_011511097.1 12 1202
XM_011512797.2 XP_011511099.1 12 2865
XM_017006363.1 XP_016861852.1 12 2839
XM_017006364.1 XP_016861853.1 12 1268
XM_017006365.1 XP_016861854.1 7 1532
GenBank mRNA AK125626.1 - 8 1218
AK128280.1 - 9 1898
AK299030.1 - 11 1277
AK312713.1 - 12 1386
BC002374.2 - 11 1257 2822859
BC003009.1 - 11 1257 2822859
BC090864.1 - 11 1239 6173605
BT006959.1 - 10 1080
CR456743.1 - 10 1080
S75295.1 - 11 1126
U20620.1 - 9 932

View Full Product Details