Gene Transcripts

Gene Symbol
Bri3
Entrez Gene ID
55950
Gene Name
brain protein I3
Gene Aliases
I3
Chromosome Location
Chr. 5: 144244437 - 144446757 on Build GRCm38
UniGene ID
Mm.38011
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001163709.1 NP_001157181.1 3 787
NM_018772.4 NP_061242.3 3 903
GenBank mRNA AK080467.1 - 3 893

View Full Product Details