Gene Transcripts

Gene Symbol
Clptm1
Entrez Gene ID
56457
Gene Name
cleft lip and palate associated transmembrane protein 1
Gene Aliases
HS9, N14
Chromosome Location
Chr. 7: 19631580 - 19665030 on Build GRCm38
UniGene ID
Mm.290960
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019649.2 NP_062623.2 7 761
GenBank mRNA AK148080.1 - 7 743
AK152410.1 - 7 751
AK152809.1 - 7 751
AK165742.1 - 6 1455
AK171901.1 - 7 752
AK181139.1 - 2 233
AK196184.1 - 2 154
BC022172.1 - 7 752 4193279
D67067.1 - 7 748

View Full Product Details