Gene Transcripts

Gene Symbol
Aqp9
Entrez Gene ID
64008
Gene Name
aquaporin 9
Gene Aliases
1700020I22Rik, AI266899, AQP-9
Chromosome Location
Chr. 9: 71110659 - 71168085 on Build GRCm38
UniGene ID
Mm.449427
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271843.1 NP_001258772.1 2 381
NM_022026.3 NP_071309.1 2 381
GenBank mRNA AB037180.1 - 2 178
AK050042.1 - 2 381
AK154151.1 - 2 385
BC024105.1 - 2 352 5096952
CT010174.1 - 2 175


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA AK050325.1 - 1437 Chr. - - [-]

View Full Product Details