Gene Transcripts

Gene Symbol
Ifitm3
Entrez Gene ID
66141
Gene Name
interferon induced transmembrane protein 3
Gene Aliases
1110004C05Rik, Cd225, Cdw217, DSPA2b, Fgls, IP15, mil-1
Chromosome Location
Chr. 7: 141009590 - 141010744 on Build GRCm38
UniGene ID
Mm.141021
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025378.2 NP_079654.1 2 454
GenBank mRNA AK003407.1 - 2 455
AK007916.1 - 2 447
AK007919.1 - 2 443
AK151890.1 - 2 420
AY082484.1 - 2 423
AY594690.1 - 2 470
BC010291.1 - 2 406 3152819

View Full Product Details