Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_025710.2 | NP_079986.1 | 2 | 674 | |
GenBank mRNA | AK003966.1 | - | 2 | 637 | |
AK012180.1 | - | 2 | 625 | ||
AK014470.1 | - | 2 | 677 | ||
AK152391.1 | - | 2 | 686 | ||
AK153139.1 | - | 2 | 661 | ||
AK180820.1 | - | 1 | 210 | ||
AK191429.1 | - | 1 | 183 | ||
AK197173.1 | - | 1 | 157 | ||
AK210928.1 | - | 1 | 159 | ||
AK212564.1 | - | 1 | 171 | ||
AK216624.1 | - | 1 | 171 | ||
AK217705.1 | - | 1 | 171 | ||
BC019934.1 | - | 2 | 681 | 5249225 | |
BC047125.1 | - | 2 | 681 |