Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_024180.5 | NP_077142.1 | 2 | 255 | |
GenBank mRNA | AK002583.1 | - | 2 | 218 | |
AK007548.1 | - | 1 | 38 | ||
AK012681.1 | - | 1 | 38 | ||
AK013455.1 | - | 2 | 228 | ||
BC002146.1 | - | 2 | 276 | 3484833 | |
DV065903.1 | - | 2 | 298 |