Gene Transcripts

Gene Symbol
Pmpca
Entrez Gene ID
66865
Gene Name
peptidase (mitochondrial processing) alpha
Gene Aliases
1200002L24Rik, 4933435E07Rik, Alpha-MPP, P-55
Chromosome Location
Chr. 2: 26389348 - 26397121 on Build GRCm38
UniGene ID
Mm.250359
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173180.3 NP_775272.1 12 1409
GenBank mRNA AK004549.1 - 12 1409
AK075965.1 - 12 1409
AK147110.1 - 12 1412
AK158990.1 - 12 1377
AK161291.1 - 12 1410
AK167975.1 - 12 1408
AK169325.1 - 12 1404
AK169342.1 - 12 1404
BC010810.1 - 12 1394 4241697

View Full Product Details